VWR® SAS Super DUO 360 Air Samplers
Supplier: VWR Collection
Certificates
About this item
When environmental contamination is very low, and the number of samples to be performed is high, operational duration can be an important factor in the choice of SAS instrument. Two sampling heads that operate simultaneously to sample 1000 litres of air, in less than 3 minutes, is the solution.
- Single body with two aspirating heads
- All manuals are available on a USB key
- Instruction manual in six different languages
The simultaneous sampling on two heads also allows you to obtain statistically representative results when using the same type of media in both heads. It is also possible to use two different media for the same sampling cycle (TSA or PCA for Total Bacterial Count and SDA for yeasts and moulds). Sampling with the two heads can halve sample process time, which is a real advantage for busy users.
Certifications: ISO 14698 and USP 1116 Pharmacopoeia.
Ordering information: For a complete system, please order the sampler you require, a battery charger, and the appropriate head.
Specifications
- Catalog No:
- 710-0867
- 710-0866
- Flow rate:
- 180 L/min
- 180 L/min
- Sampling head:
- Aluminium, stainless steel or plastic (certified ready to use)
- Aluminium, stainless steel or plastic (certified ready to use)
Specifications
Related Information
Certificates
Search by Lot NumberAvantor Cleaning and Disinfection of SAS Air Samplers.pdf Avantor Disturbance Test and Exhausted Air Test.pdf Avantor MPN Conversion Table for SAS Air Samplers.pdf Controlled Environment and Cleanroom Solutions VWR Surface Air System (SAS) Monitoring Instruments
Product Literature
More
SAS Super Air Samplers
Videos
VWR® SAS Super Air Samplers Temperature Ranges
Additional Product Details
Aspirating Heads
Product Information
Frequently Bought Together