Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Iron salicylate solution
Iron salicylate solution Reag. Ph. Eur. 1046700
Iron salicylate solution Reag. Ph. Eur. 1046700
Iron salicylate solution Reag. Ph. Eur. 1046700
Catalog # 87858.260
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Expand image
Iron salicylate solution Reag. Ph. Eur. 1046700
Catalog # 87858.260
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Specifications
Pack type:
Plastic bottle
Pk:
500 mL
Seq:
2725
Specifications
About this item
Product Details
Documentation
:
:
30492-15-0
Product Details
Documentation