Diethyl aminomalonate hydrochloride
Catalog # MOLEM86913788
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:211.65 g/mol
EINECS:236-556-8
CAS Number:13433-00-6
Formula:C₇H₁₄ClNO₄
MDL Number:MFCD00012510