Order Entry
Slovenia
Orders LinkContactUsLinkComponent
L(-)-Arabitol

L(-)-Arabitol

Glass bottle for solids, 5 g
Catalog # ROTH7738.2
Supplier:   Roth Carl
L(-)-Arabitol
Catalog # ROTH7738.2
Supplier:  Roth Carl
CAS Number:  7643-75-6
L(-)-Arabitol

Glass bottle for solids, 5 g

CAS Number:  7643-75-6
Price
80,50 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  L-(-)-Arabinitol, (2S,4S)-Pentane-1,2,3,4,5-pentaol, L-(-)-Arabitol, L-Arabinitol
  • CAS Number:7643-75-6
  • Formula:C₅H₁₂O₅
  • MDL Number:MFCD00064290
  • Molecular Weight:152.15 g/mol
  • EINECS:231-582-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.