Order Entry
Slovenia
ContactUsLinkComponent
Pedal Bins
Pedal Bins
Catalog # 129-3715
Supplier:  SEMADENI
undefined
Pedal Bins
Catalog # 129-3715
Supplier:  SEMADENI

Specifications

  • Capacity:
    6 L
  • Description:
    Pedal bin
  • Seq:
    01
  • W×D×H:
    240×260×250 mm
  • Pk:
    1

Specifications

About this item

Pedal bins are made of PP with removable inner bucket.

  • Stable with semicircular design
  • Granite colour
  • Good chemical and temperature resistance