Order Entry
Slovenia
ContactUsLinkComponent
Bovine desoxyribonuclease I Typ IV (from Pancreas)
Bovine desoxyribonuclease I Typ IV (from Pancreas)
Supplier:  Merck
Certificates
Bovine desoxyribonuclease I Typ IV (from Pancreas)
Supplier:  Merck

Specifications

  • CAS No.:
    9003-98-9
  • Source:
    Pancreas
  • Species:
    Bovine
  • Storage conditions:
    Freezer
  • Enzyme name:
    Desoxyribonuclease I, Typ IV
  • Enzyme synonyms:
    DNase I

Specifications

Frequently Bought Together