
Trimethylamine-N-oxide dihydrate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:62637-93-8
:
Formula:C₃H₉NO·2H₂O
MDL Number:MFCD00149077
:111.14 g/mol (75.11 anhy)
Melting Pt:94…98 °C
Flash Pt:95 °C
EINECS:214-675-6
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:94 to 98 °C
- Titration with HClO4:≥97.5 %
Specification Test Results
Frequently Bought Together