Potassium bis(trimethylsilyl)amide 1 mol/l in THF, Sigma-Aldrich®
Catalog # SIAL702722-100ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:M-Clarity™ quality level = MQ200
EINECS:424-100-2
CAS Number:40949-94-8
Formula:C₆H₁₈KNSi₂
MDL Number:MFCD00010330