2-Hydroxy-4'-(2-hydroxyethoxy)-2-methylpropiophenone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:106797-53-9
:
Formula:C₁₂H₁₆O₄
MDL Number:MFCD00085267
:224.2530 g/mol
:M-Clarity™ quality level = MQ200
Frequently Bought Together