Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(±)-Naringenin
(±)-Naringenin
(±)-Naringenin
ROTH8524.1
:
Roth Carl
:
(±)-Naringenin
ROTH8524.1
:
Roth Carl
:
Pack type:
Glass bottle for solids
Pk:
2,5 g
Seq:
3122
:
:
272.25 g/mol
:
207-550-2
:
480-41-1
:
93602-28-9
:
C₁₅H₁₂O₅
:
MFCD00006844