
(±)-Naringenin 97%
Catalog # ACRO139820100
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:Beige to light brown Powder
- Infrared spectrum:Conforms
- Melting point:247 to 254 °C (Dec)
- HPLC:≥96.0 %
Specification Test Results
About this item
:
:272.25 g/mol
Melting Pt:247…254 °C
EINECS:207-550-2
:GHS07
Storage Temperature:Ambient
CAS Number:480-41-1
:93602-28-9
Formula:C₁₅H₁₂O₅
MDL Number:MFCD00006844