Order Entry
Slovenia
ContactUsLinkComponent
Allantoin
Allantoin
Catalog # ROTH6327.1
Supplier:  Roth Carl
CAS Number:  
Allantoin
Catalog # ROTH6327.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g
  • Seq:
    3545
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 1-(2,5-Dioxoimidazolidin-4-yl)urea
 :
 :158.12 g/mol
EINECS:202-592-8
CAS Number:97-59-6
Formula:C₄H₆N₄O₃
MDL Number:MFCD00005260