Order Entry
Slovenia
Orders LinkContactUsLinkComponent
D(-)-Luciferin

D(-)-Luciferin

Plastic bottle for solids, 50 mg
Catalog # ROCH11626353001
Supplier:   ROCHE
D(-)-Luciferin
Catalog # ROCH11626353001
Supplier:  ROCHE
CAS Number:  2591-17-5
D(-)-Luciferin

Plastic bottle for solids, 50 mg

CAS Number:  2591-17-5
Price
1 080,00 €
Log in for availability
1 * 50 mg
Product Details & Documents
Synonyms:  D-Luciferin, (S)-2-(6-Hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid
  • CAS Number:2591-17-5
  • Formula:C₁₁H₈N₂O₃S₂
  • MDL Number:MFCD00042929
  • Molecular Weight:280.3228 g/mol
  • EINECS:219-981-3
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    50 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.