Order Entry
Slovenia
ContactUsLinkComponent
 
VWR® PBS/Tween® 20, 20X, Blot Wash Buffer, UltraPure Grade
Catalog # K875-500ML
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® PBS/Tween® 20, 20X, Blot Wash Buffer, UltraPure Grade
Catalog # K875-500ML
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    PBS/Tween®20 liquid concentrate, blot washer buffer, (20X), Ultra Pure Grade

Specifications

About this item

PBS Tween® 20 blot wash buffer is a 20X concentration with 1% Tween® 20.

  • Prevents high background by efficiently removing unbound reagents
  • Convenient liquid concentrate for blotting and ELISA applications

pH (1:20, water) @ 25 °C is 7,50 to 7,70.

Concentrate (500 ml) prepares 10 litres of a 1X buffer.

When diluted down to a 1X concentration, 0,05% Tween® 20 is present.