
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]) ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:30931-67-0
:
Formula:C₁₈H₂₄N₆O₆S₄
MDL Number:MFCD00010404
:548.68 g/mol
EINECS:250-396-6
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together