Resveratrol ≥98%
Catalog # J60790.ME
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:228.25 g/mol
Merck Index:14,8158
Melting Pt:260 °C
Density:1.36 g/cm³ (20 °C)
:GHS05
:GHS07
Storage Temperature:Freezer
CAS Number:501-36-0
Formula:C₁₄H₁₂O₃
MDL Number:MFCD00133799