Ebselen
Catalog # MOLEM88581267
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
- Environmentally Preferable:
Specifications
About this item
Synonyms: 2-Phenyl-1,2-benzoisoselenazol-3(2H)-one, 2-Phenylbenzo[d][1,2]selenazol-3(2H)-one
:
:274.20 g/mol
UN:3283
:GHS06
:GHS08
:GHS09
CAS Number:60940-34-3
Formula:C₁₃H₉NOSe
MDL Number:MFCD00210937
ADR:6.1 III