Safety boots, pull-on, Jalartic
Supplier: Jallatte
Certificates
About this item
Pull-on winter boots made from AP23™ leather (oil and water repellent). It is 10 to 15% thicker than most leathers used for safety footwear (2,4 mm).
- Warm fur for isothermal protection
- Confort-Tech integrated insole: Anatomical, antibacterial, shock absorber heel
- Softane™ dual-density PU provides excellent absorption of heel shocks and vibrations
- Perforated steel toecap (200 J)
- Puncture resistant stainless steel midsole
Colour: Honey
Certifications: EN 345-1; S3, CI
Frequently Bought Together