Ethyl methyl carbonate
Catalog # SIAL754935-50ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 mL
- Seq:2370
Specifications
About this item
:
:104.1045 g/mol
CAS Number:623-53-0
Formula:C₄H₈O₃
MDL Number:MFCD00191398