Order Entry
Slovenia
ContactUsLinkComponent
2'-Hydroxy-5'-methylacetanilide
2'-Hydroxy-5'-methylacetanilide
Catalog # MOLE11580911-5G
Supplier:  Molekula
CAS Number:  
2'-Hydroxy-5'-methylacetanilide
Catalog # MOLE11580911-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :165.19 g/mol
EINECS:228-934-6
CAS Number:6375-17-3
Formula:C₉H₁₁NO₂
MDL Number:MFCD00020183