2'-Hydroxy-5'-methylacetanilide
Catalog # MAYBSB01288.1
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:165.19 g/mol
EINECS:228-934-6
CAS Number:6375-17-3
Formula:C₉H₁₁NO₂
MDL Number:MFCD00020183