HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:148893-10-1
:
Formula:C₁₀H₁₅F₆N₆OP
MDL Number:MFCD00274639
:380.2 g/mol
:M-Clarity™ quality level = MQ200
Frequently Bought Together