Order Entry
Slovenia
ContactUsLinkComponent
Vacuum manifold, CHROMABOND® MULTI 96
Vacuum manifold, CHROMABOND® MULTI 96
Catalog # MANA738630.M
Supplier:  MACHEREY-NAGEL
undefined
Vacuum manifold, CHROMABOND® MULTI 96
Catalog # MANA738630.M
Supplier:  MACHEREY-NAGEL
Specifications
  • Description:
    CHROMABOND® MULTI 96 vacuum manifold for 96-well monoblocks with reservoir tank, 3 spacer pairs, vacuum gauge and control valve
  • Seq:
    13
  • Pk:
    1 KIT
Specifications
About this item
CHROMABOND® MULTI 96 vacuum manifold is designed for applications of CHROMABOND® MULTI 96 SPE plates as well as for CHROMAFIL® MULTI 96 filter plates.
  • Suitable for 96-well monoblocks
  • Optimum flow rate due to vacuum adjustment by control valve
  • Reservoir tank is included and further accessories are available