Order Entry
Slovenia
Orders LinkContactUsLinkComponent
HighPurity_955.jpg
Trace Metal Analysis Reagents
J.T.Baker® and Macron Fine Chemicals brand high-purity solvents and reagents give you the performance you need - minimizing the risk of contaminants that can limit accuracy while maximizing instrument sensitivity and detection power in key applications.
Product selection
Ultrapure_295.jpg
Recommended for use in your most demanding trace element analyses; analysed for up to 65 trace elements in the low ppt range...
Instra_295.jpg
Recommended for ICP-OES/AES and GFAA applications, and other applications requiring ppb trace metal testing...
AceReagent_295.jpg
Recommended for qualitative AAS applications, as well as general wet chemistry...
Macron_295.jpg
Macron Fine Chemicals acids generate highly reliable and consistent ppb trace metal analysis results...