Alcogiene SAD70 isopropyl alcohol rtu, 70% USP grade isopropyl alcohol, blended with USP grade water for injection (WFI), sterile, double bagged in 1L premium trigger sprayer 1 * 1 L
Catalog # ELITACG114IPA70WFI
Supplier: ELITE ADVANCED MATERIALS
New Product