Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PREFILLED NEUTRAL BUFFERED FORMALIN 10% 1 * 2,5 L
Catalog # ELITPNBF10201-2.5P
Supplier:
ELITE ADVANCED MATERIALS
New Product
Expand image
PREFILLED NEUTRAL BUFFERED FORMALIN 10% 1 * 2,5 L
Catalog # ELITPNBF10201-2.5P
Supplier:
ELITE ADVANCED MATERIALS
New Product