Potassium Chloride Reference Solutions A - D
Potassium chloride reference solution C, 1408,8 µS/cm (1408.8 µmho/cm) at 25 °C, 25 °C, 0,01 Molar, 1 L
Catalog # RICCR5895000-1A
Supplier: Ricca Chemical
Potassium Chloride Reference Solutions A - D
Catalog # RICCR5895000-1A
Supplier: Ricca Chemical
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:1 L
- Reference temperature:25 °C
- Description:Potassium chloride reference solution C
- Conductivity:1408,8 µS/cm (1408.8 µmho/cm) at 25 °C
- Contents:0,01 Molar



