Potassium Chloride Reference Solutions A - D
Potassium chloride reference solution A, 111,342 mS/cm (111,342 µmho/cm) at 25 °C, 25 °C, 1 Molar, 1 L
Catalog # RICCR5893000-1A
Supplier: Ricca Chemical
Potassium Chloride Reference Solutions A - D
Catalog # RICCR5893000-1A
Supplier: Ricca Chemical
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:1 L
- Reference temperature:25 °C
- Description:Potassium chloride reference solution A
- Conductivity:111,342 mS/cm (111,342 µmho/cm) at 25 °C
- Contents:1 Molar



