Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Macro Fluorescence Cuvette with PTFE Stopper, Type 21FL,17.50 ml,UV Quartz 50L×10W mm 1 * 1 items
Catalog # VWRMNP21FLUV50
Supplier:
VWR Products
Expand image
Macro Fluorescence Cuvette with PTFE Stopper, Type 21FL,17.50 ml,UV Quartz 50L×10W mm 1 * 1 items
Catalog # VWRMNP21FLUV50
Supplier:
VWR Products