Order Entry
Singapore
ContactUsLinkComponent
Canister 5 Liter, S60/61, PE-HD electrostatic conductive, UN-Y approval, dimensions (W x H x D): 165 x 241 x 195 mmNote: Canister s are marked with production date (month / year) and will normally be delivered by us within 6 months from the productio 1 * 1 items
Catalog # SCTV108329
Supplier:  S.C.A.T.
undefined
Canister 5 Liter, S60/61, PE-HD electrostatic conductive, UN-Y approval, dimensions (W x H x D): 165 x 241 x 195 mmNote: Canister s are marked with production date (month / year) and will normally be delivered by us within 6 months from the productio 1 * 1 items
Catalog # SCTV108329
Supplier:  S.C.A.T.