Order Entry
Singapore
ContactUsLinkComponent
Cleanroom TT BOPP label with release tab 8mm,1000 pcs/Roll(PE Core) Size: 4 X2 (with peforation), Corner Radius: 3mm ,Material : PMA 2074 (TMPP/RM/SRL) 1 * 1 Roll
Catalog # IBSALVWRSG20744-2
Supplier:  IBS Stock Items
Cleanroom TT BOPP label with release tab 8mm,1000 pcs/Roll(PE Core) Size: 4 X2 (with peforation), Corner Radius: 3mm ,Material : PMA 2074 (TMPP/RM/SRL) 1 * 1 Roll
Catalog # IBSALVWRSG20744-2
Supplier:  IBS Stock Items