Order Entry
Singapore
ContactUsLinkComponent
Diffuser Climet CI-302 Suitable for: Compressed Air, Nitrogen, Argon, Helium, CO, Carbon dioxide (Provided that the Particle Counter is calibrated for the specific gases) For all Climet Particle counter For all Microbial-Air-Sampler (MAS-devices) (A 1 * 1 items
Catalog # IBSACI-302
Supplier:  IBS Stock Items
Diffuser Climet CI-302 Suitable for: Compressed Air, Nitrogen, Argon, Helium, CO, Carbon dioxide (Provided that the Particle Counter is calibrated for the specific gases) For all Climet Particle counter For all Microbial-Air-Sampler (MAS-devices) (A 1 * 1 items
Catalog # IBSACI-302
Supplier:  IBS Stock Items