Order Entry
Singapore
ContactUsLinkComponent
The Extracta Plus RNA kits offer rapid purification of Total RNA from animal cells or tissues. The Extracta Plus Spin Column workflow enables simultaneous processing of multiple samples in just 25 mins to yield purified total RNA suitable for a range 1 * 1 KIT
Catalog # QUNT95214-010SMP
Supplier:  Quantabio
The Extracta Plus RNA kits offer rapid purification of Total RNA from animal cells or tissues. The Extracta Plus Spin Column workflow enables simultaneous processing of multiple samples in just 25 mins to yield purified total RNA suitable for a range 1 * 1 KIT
Catalog # QUNT95214-010SMP
Supplier:  Quantabio