QUANTOFIX® Test Strips, Chlorine dioxide
QUANTOFIX Dioxyde de chlore 3000, 0 • 10 • 25 • 50 • 100 • 250 • 500 • 1000 • 1500 • 2000 • 2500 • 3000 - mg/L, Redox reaction (potassium iodide / redox indicator), 40×40×120 mm (package), 100 test(s)
Catalog # MANA91357
Supplier: MACHEREY-NAGEL
Jump to:
Product Details & Documents
Semi-quantitative test strips for the determination of chlorine dioxide.
Chlorine dioxide - 0, 10, 25, 50, 100, 250, 500, 1000, 1500, 2000, 2500, 3000 mg/L
- Chlorine dioxide - Yellow → dark green (field 1), colourless → brown-black (field 2)
Ideal for rapid and easy analysis of chlorine dioxide directly on-site. No maintenance or calibration is required.
Specifications
Specifications
- Chemical method:Redox reaction (potassium iodide / redox indicator)
- Description:QUANTOFIX Dioxyde de chlore 3000
- Dimensions:40×40×120 mm (package)
- Tests per kit:100 test(s)
- Range:0 • 10 • 25 • 50 • 100 • 250 • 500 • 1000 • 1500 • 2000 • 2500 • 3000 - mg/L
- Cat. No.:MANA91357
- Parameters:All reagents included
- Contents:100 test strips in a tube, instruction leaflet
- Platform:Semi-quantitative test strips
- Compliance:No hazardous material
- Instrument:No
- Shelf life (months):30 (from production)
- Method:Chlorine dioxide - Redox reaction (Yellow → dark green (field 1), colorless → brown-black (field 2))
- Weight:42.8 g



