Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
UltraCenter Precision Tapered Spin ON/OFF Paddle, Solid Molded Fluoropolymer, One Piece Design, Serialized (Upper Shaft available se parately) 1 * 1 items
Catalog # QLAASPNTPD-ASTM
Supplier:
QUALITY LAB ACCESSORIES
Expand image
UltraCenter Precision Tapered Spin ON/OFF Paddle, Solid Molded Fluoropolymer, One Piece Design, Serialized (Upper Shaft available se parately) 1 * 1 items
Catalog # QLAASPNTPD-ASTM
Supplier:
QUALITY LAB ACCESSORIES