Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
10 Mesh 1.25IN (31.75mm) dia. Screen for Agilent/VanKel APP 3 Biodissolution, Stainless Steel 1 * 25 items
QLAAMSHSCR-10
:
QUALITY LAB ACCESSORIES
10 Mesh 1.25IN (31.75mm) dia. Screen for Agilent/VanKel APP 3 Biodissolution, Stainless Steel 1 * 25 items
QLAAMSHSCR-10
:
QUALITY LAB ACCESSORIES