Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Small Volume 40 Mesh Mini Basket, 316 SS, Serialized 1 * 1 items
Catalog # QLAAMINBSK-40
Supplier:
QUALITY LAB ACCESSORIES
Expand image
Small Volume 40 Mesh Mini Basket, 316 SS, Serialized 1 * 1 items
Catalog # QLAAMINBSK-40
Supplier:
QUALITY LAB ACCESSORIES