Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Model HC1000-230 Standalone Precision 1000 watt Heater/Circulator, Programmable, 230 V/ 50 Hz 1 * 1 items
Catalog # QLAAHC1000-230
Supplier:
QUALITY LAB ACCESSORIES
Expand image
Model HC1000-230 Standalone Precision 1000 watt Heater/Circulator, Programmable, 230 V/ 50 Hz 1 * 1 items
Catalog # QLAAHC1000-230
Supplier:
QUALITY LAB ACCESSORIES