Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
500mL Clear Glass Vessel with Plastic Rim and Centering Ring for use with Agilent 708-DS Baths, Serialized 1 * 1 items
Catalog # QLAAGLA5CR-70XP
Supplier:
QUALITY LAB ACCESSORIES
Expand image
500mL Clear Glass Vessel with Plastic Rim and Centering Ring for use with Agilent 708-DS Baths, Serialized 1 * 1 items
Catalog # QLAAGLA5CR-70XP
Supplier:
QUALITY LAB ACCESSORIES