Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Float-A-Lyzer Holder for Pharmatest Basket Hub, Delrin (Use for 900mL to 2000mL. 1mL, 5mL or 10mL Float-A-Lyzer) 1 * 1 items
Catalog # QLAAFLTHLD-BSKPT
Supplier:
QUALITY LAB ACCESSORIES
Expand image
Float-A-Lyzer Holder for Pharmatest Basket Hub, Delrin (Use for 900mL to 2000mL. 1mL, 5mL or 10mL Float-A-Lyzer) 1 * 1 items
Catalog # QLAAFLTHLD-BSKPT
Supplier:
QUALITY LAB ACCESSORIES