Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
45 Micron Porous Filters, UHMW Polyethylene, 1/16IN (1.6mm) ID, Hanson compatible 1 * 100 items
Catalog # QLAAFIL45S-HR-A
Supplier:
QUALITY LAB ACCESSORIES
Expand image
45 Micron Porous Filters, UHMW Polyethylene, 1/16IN (1.6mm) ID, Hanson compatible 1 * 100 items
Catalog # QLAAFIL45S-HR-A
Supplier:
QUALITY LAB ACCESSORIES