Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Glass Replacement Tubes for 3 Tube Disintegration Assembly for VanKel Testers, 38mm OD x 77.5mm L 1 * 3 items
Catalog # QLAADISTUB-03
Supplier:
QUALITY LAB ACCESSORIES
Expand image
Glass Replacement Tubes for 3 Tube Disintegration Assembly for VanKel Testers, 38mm OD x 77.5mm L 1 * 3 items
Catalog # QLAADISTUB-03
Supplier:
QUALITY LAB ACCESSORIES