Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Disintegration Basket Assembly with 3 Glass Tubes (38mm) and 10 Mesh SS Screens for VanKel Testers 1 * 1 items
Catalog # QLAADISASY-03
Supplier:
QUALITY LAB ACCESSORIES
Expand image
Disintegration Basket Assembly with 3 Glass Tubes (38mm) and 10 Mesh SS Screens for VanKel Testers 1 * 1 items
Catalog # QLAADISASY-03
Supplier:
QUALITY LAB ACCESSORIES