Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Spiral Capsule Sinker, PTFE coated Music Wire, .885IN (22.5mm) L x .575IN (14.6mm) W capacity, 8.5 coils 1 * 1 items
Catalog # QLAACAPWHT-0XS
Supplier:
QUALITY LAB ACCESSORIES
Expand image
Spiral Capsule Sinker, PTFE coated Music Wire, .885IN (22.5mm) L x .575IN (14.6mm) W capacity, 8.5 coils 1 * 1 items
Catalog # QLAACAPWHT-0XS
Supplier:
QUALITY LAB ACCESSORIES