Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Osmolality standard, 1100 mOsm/kg H 1 * 1 KIT
Catalog # REGERE-OSM-1100
Supplier:
REAGECON
Expand image
Osmolality standard, 1100 mOsm/kg H 1 * 1 KIT
Catalog # REGERE-OSM-1100
Supplier:
REAGECON