Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Tektronix Installed Option add spectrum analyzer input frequency range from 9 kHz to 3 GHz and capture bandwidth to 3 GHz 1 * 1 items
Catalog # MSPP3-SA3
Supplier:
MARKET SOURCE PART PROCESS
Expand image
Tektronix Installed Option add spectrum analyzer input frequency range from 9 kHz to 3 GHz and capture bandwidth to 3 GHz 1 * 1 items
Catalog # MSPP3-SA3
Supplier:
MARKET SOURCE PART PROCESS