Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Sample Vials, Clear Borosilicate Glass, Screw-Thread,1.9 mL,12 x 35 mm ,8-425 1 * 144 items
Catalog # QORPGLC-04878
Supplier:
QORPAK INCORPORATED
Expand image
Sample Vials, Clear Borosilicate Glass, Screw-Thread,1.9 mL,12 x 35 mm ,8-425 1 * 144 items
Catalog # QORPGLC-04878
Supplier:
QORPAK INCORPORATED