Order Entry
Singapore
ContactUsLinkComponent
CHROMABOND QuEChERS Mix II acetate extraction mix consisting of: - 4000 mg MgSO4 - 1000 mg Na acetate individually weighed and fill ed into 50 mL centrifuge tubes (PP) with screw cap (PE) pack of 50 1 * 50 items
Catalog # MANA730694.1
Supplier:  MACHEREY-NAGEL
CHROMABOND QuEChERS Mix II acetate extraction mix consisting of: - 4000 mg MgSO4 - 1000 mg Na acetate individually weighed and fill ed into 50 mL centrifuge tubes (PP) with screw cap (PE) pack of 50 1 * 50 items
Catalog # MANA730694.1
Supplier:  MACHEREY-NAGEL