Order Entry
Singapore
ContactUsLinkComponent
Standard PCD (Process Challenge Devices), for EO sterilization cycles with Mesalab BI spore strip in glassine, SGMG/6, 100 pc/bx, PCD 6.5, nominal D-value: 17 minutes 1 * 100 items
Catalog # IBSAPCD6.5
Supplier:  IBS Stock Items
Standard PCD (Process Challenge Devices), for EO sterilization cycles with Mesalab BI spore strip in glassine, SGMG/6, 100 pc/bx, PCD 6.5, nominal D-value: 17 minutes 1 * 100 items
Catalog # IBSAPCD6.5
Supplier:  IBS Stock Items