Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ultra clean bouffant cap, type: 21 , 11 gsm, white 1 * 100 items
Catalog # IBSAPP-BC-21W
Supplier:
IBS Stock Items
Expand image
ultra clean bouffant cap, type: 21 , 11 gsm, white 1 * 100 items
Catalog # IBSAPP-BC-21W
Supplier:
IBS Stock Items